missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lilolidine 98.0+%, TCI America™

$138.25 - $478.95

Chemical Identifiers

CAS 5840-01-7
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.22
MDL Number MFCD09841910
InChI Key QCCKSFHMARIKSK-UHFFFAOYSA-N
Synonym 1,7-trimethyleneindole
PubChem CID 15645629
SMILES C1CC2=CC=CC3=C2N(C1)C=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L02741G View Documents Tci America
L02741G
1g Each for $138.25
 
L02745G View Documents Tci America
L02745G
5g Each for $478.95
 
Specifications

Chemical Identifiers

5840-01-7
157.22
QCCKSFHMARIKSK-UHFFFAOYSA-N
15645629
C11H11N
MFCD09841910
1,7-trimethyleneindole
C1CC2=CC=CC3=C2N(C1)C=C3

Specifications

Yellow
1g
C11H11N
1,7-trimethyleneindole
C1CC2=CC=CC3=C2N(C1)C=C3
15645629
Crystal-Powder at 20°C
Lilolidine
86°C
5840-01-7
MFCD09841910
QCCKSFHMARIKSK-UHFFFAOYSA-N
157.22
157.22
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No