missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lignin (Alkaline), TCI America™

Manufacturer:  TCI America L0082500G

 View more versions of this product

Catalog No. L0082500G



Specifications

Specifications

Lignin (Alkaline)
500g
MFCD00147441
Black
8068-05-1
Crystal-Powder at 20°C

Chemical Identifiers

8068-05-1
MFCD00147441
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0215

TSCA : Yes

Documents