Please login to your online account to display your discounted pricing
Novus Biologicals™ Lightning-Link Rapid Type B Biotin Antibody Labeling Kit
Label antibodies, proteins, peptides and other biomolecules with biotin, using simple procedure. Set up conjugations in seconds, and use them within minutes.
$329.00 - $629.97
Specifications
Product Type Antibody Labeling Kit
Storage Requirements Store at −20°C. Avoid freeze-thaw cycles.
View More Specs
 Alert
For Research Use Only
Products
Catalog Number Mfr. No. Quantity Price Quantity    
3710010
novus biologicals™
3710010
3 x 100-200μg Reaction Each for $474.67
3710015
novus biologicals™
3710015
1 x 1-2mg Reaction Each for $629.97
3710030
novus biologicals™
3710030
Each for $329.00
Description & Specifications
Specifications
Product Type Antibody Labeling Kit
Storage Requirements Store at −20°C. Avoid freeze-thaw cycles.
  • No desalting or dialysis steps
  • Lightning-link Rapid technology can be used to label small quantities of antibody with 100% recovery
  • Type A biotin kit is intended for assays in which a streptavidin-labeled detection reagent will be used.