missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ Lightning-Link Rapid Type A Biotin Antibody Labeling Kit

Label antibodies, proteins, peptides and other biomolecules with biotin, using simple procedure. Set up conjugations in seconds, and use them within minutes.

$299.66 - $628.63

Specifications

Product Type Antibody Labeling Kit
Storage Requirements Store at −20°C. Avoid freeze-thaw cycles.
View More Specs

Alert:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
3700010 Novus Biologicals™
3700010
3 x 100-200μg Reaction Each for $516.00
3700015 Novus Biologicals$trade;
3700015
1 x 1-2mg Reaction Each for $628.63
3700030 Novus Biologicals™
3700030
3 x 10-20μg Reaction Each for $299.66
Specifications

Description

  • No desalting or dialysis steps
  • Lightning-link Rapid technology can be used to label small quantities of antibody with 100% recovery
  • Type A biotin kit is intended for assays in which a streptavidin-labeled detection reagent will be used.
Description & Specifications

Specifications

Antibody Labeling Kit
Store at −20°C. Avoid freeze-thaw cycles.