Please login to your online account to display your discounted pricing

Light's Solution, ORP Standard, Certified, LabChem™


Quantity 500mL
Packaging Amber Poly

Manufacturer: lab chem  LC161401

 View more versions of this product

Catalog No. LC161401

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 392.14
Purity Grade Certified Grade
CAS 7783-83-7
Health Hazard 1 Danger
Chemical Name or Material Light's Solution
CAS 7732-18-5
CAS 7783-85-9
CAS Max % 9.5
Color Clear, dull yellow
Quantity 500mL
CAS Max % 81.76
Packaging Amber Poly
CAS Max % 3.92
Identification Passes Test
CAS 7664-93-9
Linear Formula Fe(NH4)2(SO4)2·6H2O
Recommended Storage Room Temperature
Name Note ORP Standard
UN Number UN3264
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Physical Form Liquid
CAS Max % 4.82
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water

Specifications

Chemical Name or Material Light's Solution
CAS 7732-18-5
CAS Max % 81.76
CAS 7664-93-9
CAS Max % 9.5
CAS 7783-83-7
CAS Max % 4.82
CAS 7783-85-9
CAS Max % 3.92
Name Note ORP Standard
Purity Grade Certified Grade
Linear Formula Fe(NH4)2(SO4)2·6H2O
Formula Weight 392.14
UN Number UN3264
Accessories

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.

Room Temperature