Binder™ Light Photometry
Binder™ Light Photometry comes equipped with an individual certificate.
$4,333.46 - $4,333.46
Specifications
Includes Certificate
No. of Places 25 measuring points
Type Light photometry
View More Specs
Products 1
Catalog Number Mfr. No. Description For Use With (Application) For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Description For Use With (Application) For Use With (Equipment) Price Quantity  
08111256 View Documents Binder™
80120909
Spectral distribution of 385 to 785nm Illumination, irradiance, visible light KBF LQC 240, KBF LQC 720 Each for $4,333.46
Description
Description
  • 25 measuring points on 3 measurement levels
  • Spectral distribution of either 385 to 785nm or 250 to 785nm
  • For illumination, irradiance, visible light, and UV-A (8012-9010 only)
Specifications
Specifications
Certificate
25 measuring points
Light photometry