Thermo Scientific™ LifterSlip™ mSeries Cover Slips for Microarray Slides
Reduce the variation in hybridization fluid volume with the superior surface quality and flatness of Thermo Scientific™ LifterSlip™ mSeries Cover Slips for Microarray Slides.
Manufacturer:  Thermo Scientific™ 22X60IM5522
 View more versions of this product
Catalog No. 12-550-60
Specifications
Description
Reduce the variation in hybridization fluid volume across the microarray surface with the superior surface quality and flatness of Thermo Scientific™ LifterSlip™ mSeries Cover Slips for Microarray Slides. These slides are made of 1mm thick microarray quality, ultra flat glass.
  • Feature precise ink-printed raised edges to provide consistent dispersal and easy injection of hybridization solution
  • Manufactured without adhesives to prevent leaching at high temperatures or extended hybridization times
Description & Specifications
Specifications
mSeries LifterSlip Cover Slips for Microarray Slides
45µL