Invitrogen™ E-Gel™ Imager UV Light Base DFS Item
E-Gel Imager UV Light Base has a built-in safety mechanism that requires the E-Gel Imager Camera Hood to be affixed over the top in order to be operational
Manufacturer:  Invitrogen™ LS4466602
Catalog No. 4466602
Includes: 1 x E-Gel™ Imager UV Light Base
Description
Description
The E-Gel Imager UV Light Base is one of three interchangeable bases that work with the E-Gel Imager Camera Hood to comprise the E-Gel Imager system. The E-Gel Imager UV Light base is a compact UV light transilluminator for self-poured gels as well as E-Gel™ agarose gel cassettes stained with ethidium bromide. UV Light Base is best for ethidium bromide.
  • E-Gel Adaptor Base has been designed to provide power to either E-Gel iBase power system coupled to E-Gel Safe Imager™, or E-Gel Go Base with its integrated blue light transilluminator so E-Gel cassettes can be run right inside E-Gel Imager
  • This allows for real time image capture while samples are running on the gel
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Specifications
Specifications
E-Gel® Imager
Room Temperature
1 x E-Gel® Imager UV Light Base.
Store at room temperature.
Ethidium bromide
E-Gel® Imager
E-Gel™
SDS
Documents
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.