Eisco™ Liebig Condenser
Eisco™ Liebig Condenser is made of high quality borosilicate 3.3 glass and features an integrated inner tube and narrow jacket for efficient cooling.
$58.94 - $59.94
Specifications
Material | Borosilicate Glass |
---|---|
Type | Condenser |
For Use With (Application) | Narrow jacket for efficient cooling |
Description
- Glass Liebig condensers
- Made from high quality borosilicate glass
- Integrated inner tube
- Narrow jacket for efficient cooling
Specifications
Borosilicate Glass | |
Condenser | |
Narrow jacket for efficient cooling |