missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Labconco™ Lids for FreeZone™ Freeze Dryers
Replacement lids for FreeZone™ Freeze Dryer collector chambers.
$399.27 - $399.27
Catalog Number | Mfr. No. | For Use With (Equipment) | Material | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Material | Price | Quantity | ||||
16-108-592
|
Labconco™
7208200 |
FreeZone™ 6L, 12L, and 18L Console Freeze Dryers | Acrylic |
Each for $399.27
|
|
||||
Description
Stainless Steel, 14 in.- Stainless steel lid has 14.0 in. diameter with 3 in. diameter hole
- Optional collector lid for FreeZone™ 4.5L, 6L, 8L, 12L & 18L Freeze Dryers
- For use with solvents that attack arcylic, like acetonitrile
- Includes neoprene gasket
Specifications
Neoprene gasket |