missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lidocaine N-ethyl bromide, 99+%

Blocker of voltage-activated sodium channels

Manufacturer:  Alfa Aesar™ J60678MF

 View more versions of this product

Catalog No. AAJ60678MF



Specifications

Specifications

Lidocaine N-ethyl bromide
50mg
C16H27BrN2O
lidocaine n-ethyl bromide, qx 314 bromide, lidocaine n-ethyl bromide quaternary salt, n-2,6-dimethylphenylcarbamoylmethyl triethylammonium bromide, 2,6-dimethylphenyl carbamoyl methyl triethylazanium bromide, d0k6yk, qx-314 bromide, qx-314.br, n-ethyllidocaine bromide, lidocaine n-ethyl bromide quaternary salt
CC[N+](CC)(CC)CC(=O)NC1=C(C=CC=C1C)C.[Br-]
343.309
343.31
≥99%
White
21306-56-9
MFCD00083182
DLHMKHREUTXMCH-UHFFFAOYSA-N
[2-(2,6-dimethylanilino)-2-oxoethyl]-triethylazanium;bromide
9884487
Crystalline Solid

Chemical Identifiers

21306-56-9
343.309
DLHMKHREUTXMCH-UHFFFAOYSA-N
9884487
CC[N+](CC)(CC)CC(=O)NC1=C(C=CC=C1C)C.[Br-]
C16H27BrN2O
MFCD00083182
lidocaine n-ethyl bromide, qx 314 bromide, lidocaine n-ethyl bromide quaternary salt, n-2,6-dimethylphenylcarbamoylmethyl triethylammonium bromide, 2,6-dimethylphenyl carbamoyl methyl triethylazanium bromide, d0k6yk, qx-314 bromide, qx-314.br, n-ethyllidocaine bromide, lidocaine n-ethyl bromide quaternary salt
[2-(2,6-dimethylanilino)-2-oxoethyl]-triethylazanium;bromide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents