missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lidocaine N-ethyl bromide, 99+%

Blocker of voltage-activated sodium channels

Manufacturer:  Alfa Aesar™ J60678MC

 View more versions of this product

Catalog No. AAJ60678MC



Specifications

Specifications

White
21306-56-9
MFCD00083182
DLHMKHREUTXMCH-UHFFFAOYSA-N
[2-(2,6-dimethylanilino)-2-oxoethyl]-triethylazanium;bromide
9884487
Crystalline Solid
100mg
C16H27BrN2O
2,6-dimethylphenyl carbamoyl methyl triethylazanium bromide, d0k6yk, lidocaine n-ethyl bromide, lidocaine n-ethyl bromide quaternary salt, n-2,6-dimethylphenylcarbamoylmethyl triethylammonium bromide, n-ethyllidocaine bromide, qx 314 bromide, qx-314 bromide, qx-314.br
CC[N+](CC)(CC)CC(=O)NC1=C(C=CC=C1C)C.[Br-]
343.309
343.31
≥99%

Chemical Identifiers

21306-56-9
343.309
DLHMKHREUTXMCH-UHFFFAOYSA-N
9884487
CC[N+](CC)(CC)CC(=O)NC1=C(C=CC=C1C)C.[Br-]
C16H27BrN2O
MFCD00083182
2,6-dimethylphenyl carbamoyl methyl triethylazanium bromide, d0k6yk, lidocaine n-ethyl bromide, lidocaine n-ethyl bromide quaternary salt, n-2,6-dimethylphenylcarbamoylmethyl triethylammonium bromide, n-ethyllidocaine bromide, qx 314 bromide, qx-314 bromide, qx-314.br
[2-(2,6-dimethylanilino)-2-oxoethyl]-triethylazanium;bromide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

Documents