MilliporeSigma™ LiChroCART™ Column/LiChrospher™ RP-18 ADS Sorbent HPLC Cartridges, 25μm
LiChroCART column prepacked w/LiChrospher RP-18 ADS sorbent
$3,387.22 - $4,466.37
Specifications
Length (Metric) 25 mm
Quantity 3/Pk.
Type LiChroCART Column/LiChrospher RP-18 ADS Sorbent HPLC Cartridge
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
M1503860001
View Documents
MilliporeSigma™
1503860001
2 mm
Each for $4,466.37
M1509470001
View Documents
MilliporeSigma™
1509470001
4 mm
Each for $3,387.22
Description
Description
LiChrospher™ ADS allows the fully automated, LC integrated sample preparation of hydrophobic, low molecular weight analytes from untreated samples like serum, milk, fermentation broth of food homogenates. The enrichment is based on two chromatographic processes: reversed phase and size exclusion chromatography since LiChrospher™ ADS sorbents are spherical silica gel particles (25μm) with DIOL modification on the exterior surface and alkyl modifications on the surface of the interior pores (ID: 6nm). Depending on the hydrophobicity of the analyte three types of precolumns are available: C-4, C-8 and C-18.
Specifications
Specifications
25 mm
3/Pk.
LiChroCART Column/LiChrospher RP-18 ADS Sorbent HPLC Cartridge
SDS