Please login to your online account to display your discounted pricing

Liarozole hydrochloride, Tocris

Blocks retinoic acid metabolism

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material Liarozole hydrochloride
Assay Percent Range >99%
Molecular Formula C17H13ClN4·HCl
Formula Weight 345.23
Synonym 5-[(3-Chlorophenyl)-1H-imidazol-1-ylmethyl]-1H-benzimidazole hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

270510

 
r & d systems
RDS270510
10mg Each for $225.77

270550

 
r & d systems
RDS270550
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 345.23
Quantity 10mg
Chemical Name or Material Liarozole hydrochloride
Molecular Formula C17H13ClN4·HCl
CAS 145858-50-0
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >99%
Synonym 5-[(3-Chlorophenyl)-1H-imidazol-1-ylmethyl]-1H-benzimidazole hydrochloride
Safety and Handling

Desiccate at Room Temperature