missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ LH (Luteinizing Hormone), Mouse Monoclonal Antibody

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ LH (Luteinizing Hormone), Mouse Monoclonal Antibody provides accurate, reproducible results.

$356.39 - $976.47

Specifications

Antigen LH (Luteinizing Hormone)
Clone SPM103
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MS9078P Thermo Scientific™ Lab Vision™
MS9078P
1mL Each for $976.47
MS9078P1 Thermo Scientific™ Lab Vision™
MS9078P1
500μL Each for $385.35
MS9078R7 Thermo Scientific™ Lab Vision™
MS9078R7
7mL Each for $356.39
Specifications

Description

Ideal for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ LH (Luteinizing Hormone), Mouse Monoclonal Antibody provides accurate, reproducible results.

LH is a tropic hormone, which modulates the secretory activity of other endocrine glands. It is produced in the anterior hypophysis of the pituitary gland. The glycoprotein hormone, LH, like follicle stimulating hormone and thyroid stimulating hormone, is composed of a common alpha-sububit but also a specific beta-subunit, which characterizes each of these hormones.

Host Species: Mouse

Clone: SPM103

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Immunogen: LH purified from human pituitary gland

Molecular Weight: 14kDa (β), 28kDa (α/β)

Positive Control: Anterior pituitary

Cellular Localization: Cytoplasmic. This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

LH (Luteinizing Hormone)
Immunohistochemistry (Paraffin)
Unlabeled
Murine
Steroids Hormones and Growth Factors
LH purified from human pituitary gland
SPM103
Monoclonal
Human
RUO
Primary
IgG1 kappa