Please login to your online account to display your discounted pricing

LH 846, Tocris™

Selective casein kinase 1δ inhibitor

$208.25 - $867.95

Chemical Identifiers

Chemical Name or Material LH 846
Assay Percent Range >98%
Molecular Formula C16H13ClN2OS
Formula Weight 316.81
Synonym N-(5-Chloro-6-methyl-2-benzothiazolyl)benzeneacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

489610

 
r & d systems
RDS489610
10mg Each for $208.25

489650

 
r & d systems
RDS489650
50mg Each for $867.95
Description & Specifications

Specifications

Formula Weight 316.81
Quantity 10mg
Molecular Formula C16H13ClN2OS
Synonym N-(5-Chloro-6-methyl-2-benzothiazolyl)benzeneacetamide
CAS 639052-78-1
Chemical Name or Material LH 846
Assay Percent Range >98%
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C