Please login to your online account to display your discounted pricing

LG 101506, Tocris™

Selective RXR modulator

$259.29 - $1,075.94

Chemical Identifiers

Chemical Name or Material LG 101506
Assay Percent Range >98%
Molecular Formula C25H34F2O3
Formula Weight 420.53
Synonym (2E,4E,6Z)-7-[2-(2,2-Difluoroethoxy)-3,5-bis(1,1-dimethylethyl)phenyl]-3-methyl-2,4,6-octatrienoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

350850

 
r & d systems
RDS350850
50mg Each for $1,075.94

350810

 
r & d systems
RDS350810
10mg Each for $259.29
Description & Specifications

Specifications

Formula Weight 420.53
Quantity 50mg
CAS 331248-11-4
Synonym (2E,4E,6Z)-7-[2-(2,2-Difluoroethoxy)-3,5-bis(1,1-dimethylethyl)phenyl]-3-methyl-2,4,6-octatrienoic acid
Chemical Name or Material LG 101506
Assay Percent Range >98%
Molecular Formula C25H34F2O3
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C