Please login to your online account to display your discounted pricing

LG 100754, Tocris™

RXR:PPAR agonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material LG 100754
Assay Percent Range >99%
Molecular Formula C26H36O3
Formula Weight 396.56
Synonym (2E,4E,6Z)-3-Methyl-7-(5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-propoxy-3-naphthalenyl)-2,4,6-octatrienoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

383110

 
r & d systems
RDS383110
10mg Each for $252.39

383150

 
r & d systems
RDS383150
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 396.56
Quantity 10mg
Chemical Name or Material LG 100754
Synonym (2E,4E,6Z)-3-Methyl-7-(5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-propoxy-3-naphthalenyl)-2,4,6-octatrienoic acid
Assay Percent Range >99%
Molecular Formula C26H36O3
CAS 180713-37-5
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C