Please login to your online account to display your discounted pricing

LFM-A13, Tocris


Potent, selective BTK inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS130010

 View more versions of this product

Catalog No. 130010

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material LFM-A13
Quantity 10mg
CAS 62004-35-7
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C11H8Br2N2O2
Formula Weight 360
Synonym 2-Cyano-N-(2, 5-dibromophenyl)-3-hydroxy-2-butenamide

Specifications

Chemical Name or Material LFM-A13
Assay Percent Range >98%
Molecular Formula C11H8Br2N2O2
Formula Weight 360
Synonym 2-Cyano-N-(2, 5-dibromophenyl)-3-hydroxy-2-butenamide
Accessories

Store at Room Temperature