Please login to your online account to display your discounted pricing

LFM-A13, Tocris

Potent, selective BTK inhibitor

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material LFM-A13
Assay Percent Range >99%
Molecular Formula C11H8Br2N2O2
Formula Weight 360
Synonym 2-Cyano-N-(2, 5-dibromophenyl)-3-hydroxy-2-butenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

130010

 
r & d systems
RDS130010
10mg Each for $221.01

130050

 
r & d systems
RDS130050
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 360
Quantity 10mg
Molecular Formula C11H8Br2N2O2
Chemical Name or Material LFM-A13
Synonym 2-Cyano-N-(2, 5-dibromophenyl)-3-hydroxy-2-butenamide
CAS 62004-35-7
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature