missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

LFM-A13, Tocris

Potent, selective BTK inhibitor

Manufacturer:  R & D Systems RDS130050

 View more versions of this product

Catalog No. 130050


 

Specifications Description & Specifications

Specifications

LFM-A13
62004-35-7
C11H8Br2N2O2
2-Cyano-N-(2, 5-dibromophenyl)-3-hydroxy-2-butenamide
50mg
>98%
360

Chemical Identifiers

>98%
360
LFM-A13
C11H8Br2N2O2
2-Cyano-N-(2, 5-dibromophenyl)-3-hydroxy-2-butenamide

Safety and Handling

Recommended Storage : Store at Room Temperature