Learn More
Levulinic Acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 125140010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Levulinic acid | |
33.0°C to 35.0°C | |
1.1300g/mL | |
98°C | |
98+% | |
C5H8O3 | |
CH3COCH2CH2CO2H | |
1 kg | |
01,564; 02,239; 10,230 | |
15, 5526 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol and ether,insoluble in aliphatic hydrocarbons | |
CC(=O)CCC(=O)O | |
116.12 | |
CHEBI:45630 | |
98+% |
123-76-2 | |
Yellow | |
245.0°C to 246.0°C | |
Authentic | |
Glass bottle | |
1.4396 to 1.4426 | |
MFCD00002796 | |
03, 671 | |
1.13 | |
levulinic acid, laevulinic acid, pentanoic acid, 4-oxo, levulic acid, 4-oxovaleric acid, 3-acetylpropionic acid, 4-ketovaleric acid, leva, gamma-ketovaleric acid, acetopropionic acid | |
JOOXCMJARBKPKM-UHFFFAOYSA-N | |
4-oxopentanoic acid | |
11579 | |
116.12 | |
Liquid After Melting |
Chemical Identifiers
123-76-2 | |
116.12 | |
JOOXCMJARBKPKM-UHFFFAOYSA-N | |
11579 | |
4-oxopentanoic acid |
C5H8O3 | |
MFCD00002796 | |
levulinic acid, laevulinic acid, pentanoic acid, 4-oxo, levulic acid, 4-oxovaleric acid, 3-acetylpropionic acid, 4-ketovaleric acid, leva, gamma-ketovaleric acid, acetopropionic acid | |
CHEBI:45630 | |
CC(=O)CCC(=O)O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen
GHS Signal Word: Warning
EINECSNumber : 204-649-2
RUO – Research Use Only