missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Levocetirizine Dihydrochloride, TCI America™

Manufacturer:  TCI America L02645G

 View more versions of this product

Catalog No. L02645G



Specifications

Specifications

Levocetirizine Dihydrochloride
214°C
130018-87-0
MFCD07366507
PGLIUCLTXOYQMV-GHVWMZMZSA-N
2-[2-[4-[(R)-(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride
9955977
Crystal-Powder at 20°C
White
5g
C21H27Cl3N2O3
2-[2-[4-[(R)-(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride
C1CN(CCN1CCOCC(=O)O)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl.Cl.Cl
461.808
461.81
≥98.0% (HPLC,T)

Chemical Identifiers

130018-87-0
461.808
PGLIUCLTXOYQMV-GHVWMZMZSA-N
9955977
C1CN(CCN1CCOCC(=O)O)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl.Cl.Cl
C21H27Cl3N2O3
MFCD07366507
2-[2-[4-[(R)-(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride
2-[2-[4-[(R)-(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : AG0990000

TSCA : No

Documents