missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Levamisole Hydrochloride [for Biochemical Research], TCI America™

Manufacturer:  TCI America L02315G

 View more versions of this product

Catalog No. L02315G



Specifications

Specifications

Levamisole Hydrochloride [for Biochemical Research]
230°C
16595-80-5
MFCD00012675
levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris
C1CSC2=NC(CN21)C3=CC=CC=C3.Cl
240.749
CHEBI:6433
Crystal-Powder at 20°C
White
5g
C11H13ClN2S
2811
LAZPBGZRMVRFKY-HNCPQSOCSA-N
(6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride
27944
240.75
≥99.0% (T)

Chemical Identifiers

16595-80-5
240.749
LAZPBGZRMVRFKY-HNCPQSOCSA-N
27944
(6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride
C11H13ClN2S
MFCD00012675
levamisole hydrochloride, levamisole hcl, ergamisol, tramisol, tramisole, decaris, --tetramisole hydrochloride, niratic hydrochloride, ascaridil, dekaris
CHEBI:6433
C1CSC2=NC(CN21)C3=CC=CC=C3.Cl
Safety and Handling

Safety and Handling

RTECSNumber : NJ5960000

TSCA : No

Documents