Please login to your online account to display your discounted pricing

Alfa Aesar™ Leuprolide, Human, Synthetic

For cellular and molecular biology applications

Specifications

Product Type Leuprolide, human, synthetic
Molecular Weight 1209.41
Sequence [Des-Gly10, D-Leu6, Pro-NHEt9]LH-RH
Format Lyophilized powder
Common Name Leuprorelin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ62967LB0

 
alfa aesar™
J62967LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Leuprolide, human, synthetic
Molecular Weight 1209.41
Sequence [Des-Gly10, D-Leu6, Pro-NHEt9]LH-RH
Format Lyophilized powder
Common Name Leuprorelin
Regulatory Status RUO
Purification Method HPLC