missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Leukadherin 1, Tocris™

Allosteric activator of CD11b/CD18

$142.08 - $598.33

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C22H15NO4S2
Formula Weight 421.49
Synonym 4-[(5-[[4-Oxo-3-(phenylmethyl)-2-thioxo-5-thiazolidinylidene]methyl]-2-furanyl]benzoic acid
Chemical Name or Material Leukadherin 1
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
492150 R & D Systems
RDS492150
50mg Each for $598.33
492110 R & D Systems
RDS492110
10mg Each for $142.08
Specifications Description & Specifications

Chemical Identifiers

>98%
421.49
Leukadherin 1
C22H15NO4S2
4-[(5-[[4-Oxo-3-(phenylmethyl)-2-thioxo-5-thiazolidinylidene]methyl]-2-furanyl]benzoic acid

Specifications

344897-95-6
C22H15NO4S2
4-[(5-[[4-Oxo-3-(phenylmethyl)-2-thioxo-5-thiazolidinylidene]methyl]-2-furanyl]benzoic acid
Leukadherin 1
>98%
421.49
50mg

Safety and Handling

Recommended Storage : Store at +4°C