Please login to your online account to display your discounted pricing

Alfa Aesar™ [Leu&superscript_3;&superscript_1;, Pro&superscript_3;&superscript_4;] Peptide YY, Human

For cellular and molecular biology applications

Specifications

Product Type [Leu31, Pro34] Peptide YY, human
Molecular Weight 4292.76
Storage Requirements -30° to -10°C
Sequence PYY, human
Format Powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66148EXD

 
alfa aesar™
J66148EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66148MCR

 
alfa aesar™
J66148MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Leu31, Pro34] Peptide YY, human
Molecular Weight 4292.76
Storage Requirements -30° to -10°C
Sequence PYY, human
Format Powder
Regulatory Status RUO
Purification Method HPLC