Please login to your online account to display your discounted pricing

Alfa Aesar™ [Leu&superscript_3;&superscript_1;, Pro&superscript_3;&superscript_4;] Neuropeptide Y (1-36), Porcine


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66699EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66699EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 0.5mg
Format Solid
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type [Leu31, Pro34] Neuropeptide Y (1-36), porcine
Molecular Weight 4222.69
Purification Method HPLC
Cross Reactivity Porcine