missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals [Leu13] Motilin, Human, Porcine
For cellular and molecular biology applications
$325.45 - $325.45
Specifications
Species | Human, Porcine |
---|---|
Purification Method | HPLC |
Content And Storage | -30°C to -10°C |
Format | Solid |
Molecular Weight (g/mol) | 2681.10 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ66962EXD
|
Thermo Scientific Chemicals
J66962EXD |
0.5 mg |
Each for $325.45
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Human, Porcine | |
-30°C to -10°C | |
2681.10 | |
RUO - research use only |
HPLC | |
Solid | |
[Leu13] Motilin, human, porcine | |
Phe-Val-Pro-Ile-Phe-Thr-Tyr-Gly-Glu-Leu-Gln-Arg-Leu-Gln-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln |
RUO – Research Use Only