Please login to your online account to display your discounted pricing

Alfa Aesar™ [Leu&superscript_1;&superscript_3;] Motilin, Human, Porcine


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66962EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66962EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 0.5mg
Format Solid
Sequence Phe-Val-Pro-Ile-Phe-Thr-Tyr-Gly-Glu-Leu-Gln-Arg-Leu-Gln-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type [Leu13] Motilin, human, porcine
Molecular Weight 2681.10
Purification Method HPLC
Cross Reactivity Human, Porcine