Please login to your online account to display your discounted pricing

Alfa Aesar™ Leu-Enkephalin, Certified Mass Spec Grade

For cellular and molecular biology applications

Specifications

Product Type Leu-Enkephalin, Certified Mass Spec Grade
Molecular Weight 555.40
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence YGGFL
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67650LPL

 
alfa aesar™
J67650LPL
1 vial, 500 pmol CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Leu-Enkephalin, Certified Mass Spec Grade
Molecular Weight 555.40
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence YGGFL
Regulatory Status RUO
Purification Method HPLC