Please login to your online account to display your discounted pricing

Alfa Aesar™ Leu-Enkephalin, Certified Mass Spec Grade

For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67650-ODL

 View all options for this product

Catalog No. AAJ67650ODL

 Please call Customer Service at 1-800-234-7437 or send an email to for assistance.

Description & Specifications


Molecular Weight 555.40
Purification Method HPLC
Quantity 3 vials, 500 pmol/vial
Format Lyophilized powder
Sequence YGGFL
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Leu-Enkephalin, Certified Mass Spec Grade