Please login to your online account to display your discounted pricing

Alfa Aesar™ Leptomycin B, ≥99%, 1mM soln. in ethanol

Chemical Identifiers

Chemical Name or Material Leptomycin B
CAS 87081-35-4
Formula Weight 540.73
MDL Number MFCD06795848
UN Number UN1993
Merck Index 14,5444
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ63784EXG

  SDS
alfa aesar™
J63784EXG
100μg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 540.73
Chemical Name or Material Leptomycin B
Assay ≥99%
MDL Number MFCD06795848
CAS 87081-35-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Physical Form Liquid
Odor Alcohol-like
UN Number UN1993
Merck Index 14,5444
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Clear colorless
Storage Note 1 -30° to -10°C
Quantity 100μg
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Concentration 1mM soln. in ethanol

  • Potent, specific inhibitor of nuclear export signal (NES)-dependent protein export from the nucleus

Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapor.

-30° to -10°C