Leica Microsystems Inoculating Loops and Holders
Loops and holders designed for culturing, transferring, streaking or isolating tissue cultures in cytology or histology
Supplier: Leica Microsystems 39105105
Description
- Holders constructed of durable, heat-resistant brass with insulating sleeve
- Loops allow an inoculum to be taken from a liquid by using surface tension
Specifications
8 in. | |
Brass | |
20.3 cm | |
Inoculating Loop Holder |