missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lecithin, 60%, egg

$108.09 - $740.22

Chemical Identifiers

CAS 8002-43-5
Molecular Formula C42H80NO8P
Molecular Weight (g/mol) 758.075
MDL Number MFCD00131449
InChI Key JLPULHDHAOZNQI-JLOPVYAASA-N
Synonym 3-sn-phosphatidylcholine, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, l-, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution
PubChem CID 16213884
ChEBI CHEBI:86658
IUPAC Name [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6057614 View Documents Alfa Aesar™
J6057614
25g Each for $108.09
 
AAJ6057622 View Documents Alfa Aesar™
J6057622
100g Each for $336.18
 
AAJ6057630 View Documents Alfa Aesar™
J6057630
250g Each for $740.22
 
Description

Description

Applications
Lecithin is used as an emulsifying and stabilizing agent in food, pharmaceutical and cosmetic industries. Lecithin is taken as a medicine and is also used in the manufacture of medicines. Soybean lecithin is useful for its application in liquid ram semen. Fractionation process of sunflower lecithin with absolute ethanol was used for obtaining enriched fractions in certain phospholipids.

Solubility
Soluble in diethyl ether, absolute alcohol, chloroform and petroleum ether. Sparingly soluble in benzene. Insoluble in water and acetone .

Notes
Light, heat, moisture and air-sensitive. Do not store in direct sunlight. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

8002-43-5
758.075
JLPULHDHAOZNQI-JLOPVYAASA-N
16213884
[(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
C42H80NO8P
MFCD00131449
3-sn-phosphatidylcholine, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, l-, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution
CHEBI:86658
CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

Odorless
25g
C42H80NO8P
5209585
3-sn-phosphatidylcholine, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, l-, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution
CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
758.075
CHEBI:86658
60%
Brown to Yellow
8002-43-5
MFCD00131449
14,5428
JLPULHDHAOZNQI-JLOPVYAASA-N
[(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
16213884
Powder
Lecithin, egg
Safety and Handling

Safety and Handling

RTECSNumber : OG7565000

TSCA : TSCA

Recommended Storage : −30° to −10°C