Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead zirconium titanium oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.9% (metals basis)

Magnetron sputtering source

$1,125.55 - $1,125.55

Chemical Identifiers

Chemical Name or Material Lead zirconium titanium oxide sputtering target
CAS 12626-81-2
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00212270
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41068KS

  SDS
alfa aesar™
41068KS
1 each Each for $1,125.55
Description & Specifications

Specifications

Chemical Name or Material Lead zirconium titanium oxide sputtering target
TSCA TSCA
Odor Odorless
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00212270
CAS 12626-81-2
Color Silvery white
Quantity 1 each
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.