missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead wire, 1.0mm (0.04 in.) dia., 99.9% (metals basis), Alfa Aesar™

Form: approx. 8.9g/m

Manufacturer:  Thermo Fisher Scientific 12605H4

 View more versions of this product

Catalog No. AA12605H4



Specifications

Specifications

Lead
7439-92-1
99.9% (Metals basis)
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
lead
207.2
CHEBI:27889
Gray
100m
Gray wire
Pb
blei
[Pb]
Odorless
5352425
Wire, ≈8.9g/m

Chemical Identifiers

7439-92-1
207.2
WABPQHHGFIMREM-UHFFFAOYSA-N
5352425
lead
Pb
MFCD00134050
blei
CHEBI:27889
[Pb]
Safety and Handling

Safety and Handling

P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c

H350-H360FD-H362-H372

EINECSNumber : 231-100-4

RTECSNumber : OF7525000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents