missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead Wire, 0.5mm (0.02 in.) dia., 99.998% (metals basis), Alfa Aesar™, Puratronic™

Form: approx. 2.23g/m

Manufacturer:  Alfa Aesar 11902G5

 View more versions of this product

Catalog No. AA11902G5



Specifications

Specifications

Lead
7439-92-1
99.998% (Metals basis)
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
lead
207.2
CHEBI:27889
Gray
5m
Gray wire
Pb
blei
[Pb]
Odorless
5352425
Wire, 0.5mm (0.02 in.) dia.

Chemical Identifiers

7439-92-1
207.2
WABPQHHGFIMREM-UHFFFAOYSA-N
5352425
lead
Pb
MFCD00134050
blei
CHEBI:27889
[Pb]
Safety and Handling

Safety and Handling

H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

EINECSNumber : 231-100-4

Documents