Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead titanium oxide, polymeric precursor, oxide >30 wt%

Used in electronic devices

Chemical Identifiers

Chemical Name or Material Lead titanium oxide
CAS 12060-00-3
Name Note Polymeric precursor
MDL Number MFCD00011159
UN Number UN1993
Synonym Lead titanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3975604

  SDS
alfa aesar™
3975604
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3975609

  SDS
alfa aesar™
3975609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3975618

  SDS
alfa aesar™
3975618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3975630

  SDS
alfa aesar™
3975630
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Viscous Liquid
Chemical Name or Material Lead titanium oxide
Quantity 2g
CAS 12060-00-3
Synonym Lead titanate
Name Note Polymeric precursor
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Health Hazard 1 Danger
Concentration Oxide >30 wt%
UN Number UN1993
Health Hazard 2 GHS H Statement
H225-H360-H373-H302-H332-H319
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 12° (54°F)
Color Brown
Solubility Information Soluble in alcohols
Sensitivity Moisture Sensitive
MDL Number MFCD00011159
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H360-H373-H302-H332-H319
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.