Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead Tin, solder alloy, 3.18mm (0.125 in.) dia.

Chemical Identifiers

Chemical Name or Material Lead Tin, Solder Alloy
Name Note 3.18mm (0.125 in.) dia.
MDL Number MFCD00798549
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41025TB

 
alfa aesar™
41025TB
113g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4102529

 
alfa aesar™
4102529
225g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Lead Tin, Solder Alloy
TSCA TSCA
Name Note 3.18mm (0.125 in.) dia.
MDL Number MFCD00798549
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Avg. Mol. Wt. or Mol. Wt. Range Pb:Sn; 70:30 wt%
Quantity 113g

  • For use with copper, copper alloys, lead, steel, and nickel alloys
  • Recommended for machine and torch soldering

Safety and Handling

Danger

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.