Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead tin oxide dihydrate

Chemical Identifiers

Chemical Name or Material Lead tin oxide dihydrate
CAS 12036-31-6
Linear Formula PbSnO3·2H2O
Formula Weight 409.94 (373.91 Anhydrous)
MDL Number MFCD00049760
UN Number UN3077
Synonym Lead stannate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2266014

 
alfa aesar™
2266014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2266022

 
alfa aesar™
2266022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 409.94 (373.91 Anhydrous)
Chemical Name or Material Lead tin oxide dihydrate
TSCA TSCA
Odor Odorless
Physical Form Powder
Linear Formula PbSnO3·2H2O
MDL Number MFCD00049760
CAS 12036-31-6
UN Number UN3077
Synonym Lead stannate
Color White
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.