Please login to your online account to display your discounted pricing

Lead subacetate, ACROS Organics™


Quantity 1kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  217540010

 View more versions of this product

Catalog No. AC217540010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥72% (Pb)
Purity Grade Extra Pure
Physical Form Granular Powder
Formula Weight 807.69
Quantity 1kg
Color White
Merck Index 15, 5474
RTECS Number OF8750000
EINECS Number 215-630-3
TSCA TSCA
Infrared Spectrum Authentic
Nitrate 200ppm max.
Copper (Cu) 200ppm max.
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
May damage the unborn child. Suspected of damaging fertility.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note c.p.
MDL Number MFCD00013041
CAS 1335-32-6
Health Hazard 1 Danger
Chemical Name or Material Lead subacetate
Molecular Formula C4H10O8Pb3
Linear Formula (CH3CO2)2Pb·2Pb(OH)2
Insoluble Matter 0.05% max. (in dilute acetic acid)
Iron (Fe) 200ppm max.
Packaging Plastic bottle
Synonym Basic lead acetate
Chlorine (Cl) 300ppm max.

Specifications

Chemical Name or Material Lead subacetate
Name Note c.p.
CAS 1335-32-6
Purity Grade Extra Pure
Assay ≥72% (Pb)
Molecular Formula C4H10O8Pb3
Linear Formula (CH3CO2)2Pb·2Pb(OH)2
Formula Weight 807.69
MDL Number MFCD00013041
Synonym Basic lead acetate
Accessories

Danger

GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
May damage the unborn child. Suspected of damaging fertility.

SDS & Safety Documents