Please login to your online account to display your discounted pricing

Lead Standard Solution (0.1 per cent Pb) R, Ricca Chemical

$279.45 - $279.45

Chemical Identifiers

Chemical Name or Material Lead Standard Solution
Name Note Reagent
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 10099-74-8
CAS Min % 0.16
CAS Max % 0.16
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

43028

 
ricca chemical company
RI43028
Poly natural 250mL Each for $279.45

430216

  SDS
ricca chemical company
RI430216
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Lead Standard Solution
Name Note Reagent
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 10099-74-8
CAS Min % 0.16
CAS Max % 0.16
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 0.1% Pb
Color Clear colorless
Quantity 250mL