missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lead Standard, 10 ppm Pb, Ricca Chemical
Supplier: Ricca Chemical Company R4297000100A
Specifications
Lead Standard | |
Natural Poly Bottle | |
97.0,1.29,0.05,0.0010 | |
Colorless | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
lead | |
5352425 |
7439-92-1,7697-37-2,7697-37-2,7732-18-5 | |
100 mL | |
100.0,1.35,0.05,0.0010 | |
Pb | |
[Pb] | |
207.2 | |
CHEBI:27889 |