Please login to your online account to display your discounted pricing

Lead Standard, 10ppm Pb, Ricca Chemical


Quantity 500mL
Packaging Plastic Bottle

Manufacturer: ricca chemical company  RIR4297000500A

 View more versions of this product

Catalog No. R4297000500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Clear colorless
Physical Form Liquid
Quantity 500mL
Chemical Name or Material Lead Standard
CAS 7732-18-5
CAS Max % 0.05
CAS 7439-92-1
CAS Min % 0.0010
CAS Max % 0.0010
CAS Min % 97.0
CAS Max % 100.0
CAS 7697-37-2
Packaging Plastic Bottle
CAS Min % 1.29
CAS Max % 1.35
CAS 7697-37-2
CAS Min % 0.05

Specifications

Chemical Name or Material Lead Standard
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 7697-37-2
CAS Min % 1.29
CAS Max % 1.35
CAS 7697-37-2
CAS Min % 0.05
CAS Max % 0.05
CAS 7439-92-1
CAS Min % 0.0010
CAS Max % 0.0010
Physical Form Liquid