Please login to your online account to display your discounted pricing

Lead Standard, 10,000μg/mL in 2-5% HNO3 (Plasma Grade), SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$267.83 - $504.43

Chemical Identifiers

Chemical Name or Material Lead Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
View More Specs
Includes:  NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PLPB2-3Y

 
spex certiprep llc
PLPB23Y
Triple-leached LDPE bottle 125mL Each for $267.83

PLPB2-3X

 
spex certiprep llc
PLPB23X
Triple-leached LDPE bottle 500mL Each for $504.43
Description & Specifications

Specifications

Quantity 125mL
Concentration or Composition (by Analyte or Components) Matrix&tab;Concentration&tab;Cat. No.
2-5% HNO3&tab;10, 000μg/mL&tab;PLPB2-3Y, -3X
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Health Hazard 1 Causes skin irritation, serious eye irritation. May cause respiratory irritation
Packaging Triple-leached LDPE bottle
Recommended Storage Ambient
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
DOT Information Hazardous
Solvent or Matrix 5% HNO3
Purity Grade Plasma Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Chemical Name or Material Lead Standard

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Pb10,000μg/mL

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Causes skin irritation, serious eye irritation. May cause respiratory irritation

Ambient