Please login to your online account to display your discounted pricing

Lead Standard, 1,000ug/mL in 2-5% HNO3 (Plasma Grade), SPEX CertiPrep™

$81.31 - $245.34

Chemical Identifiers

Chemical Name or Material Lead Standard
CAS 7439-92-1
CAS Min % 1.0
CAS Max % 1.0
CAS 7697-37-2
CAS Min % 5.0
CAS Max % 5.0
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 94.0
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PLPB22T

 
spex certiprep llc
PLPB22T
Poly Bottle 250mL Each for $245.34

PLPB22M

 
spex certiprep llc
SPXPLPB22M
Triple-leached LDPE bottle 30mL Each for $81.31
Description & Specifications

Specifications

Quantity 250mL
Health Hazard 1 Causes skin irritation, serious eye irritation
Recommended Storage Ambient
DOT Information Nonhazardous
Concentration or Composition (by Analyte or Components) Matrix&tab;Concentration&tab;Cat. No.
2-5% HNO3&tab;1000μg/mL&tab;PLPB2-2Y, -2T, -2X
Purity Grade Plasma Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Chemical Name or Material Lead Standard
CAS Max % 5.0
CAS Min % 5.0
CAS Min % 1.0
CAS 7732-18-5
CAS Max % 94.0
CAS Max % 1.0
CAS 7697-37-2
CAS Min % 94.0
Packaging Poly Bottle
CAS 7439-92-1

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Causes skin irritation, serious eye irritation

Ambient