Please login to your online account to display your discounted pricing

Lead Standard, 1,000ug/mL in 2-5% HNO3 (Plasma Grade), SPEX CertiPrep™


Quantity 30mL
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  SPXPLPB22M

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Catalog No. PLPB22M

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 30mL
Concentration or Composition (by Analyte or Components) Matrix&tab;Concentration&tab;Cat. No.
2-5% HNO3&tab;1000μg/mL&tab;PLPB2-2Y, -2T, -2X
CAS Min % 94.0
CAS 7732-18-5
CAS 7697-37-2
CAS Min % 5.0
Purity Grade Plasma Grade
CAS Max % 94.0
CAS Min % 1.0
CAS 7439-92-1
CAS Max % 1.0
CAS Max % 5.0
Health Hazard 1 Causes skin irritation, serious eye irritation
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
DOT Information Nonhazardous
Solvent or Matrix 2% HNO3
Packaging Triple-leached LDPE bottle
Chemical Name or Material Lead Standard
Recommended Storage Ambient

Specifications

Chemical Name or Material Lead Standard
CAS 7439-92-1
CAS Min % 1.0
CAS Max % 1.0
CAS 7697-37-2
CAS Min % 5.0
CAS Max % 5.0
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 94.0
Purity Grade Plasma Grade

AnalyteConcentration
Pb1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Causes skin irritation, serious eye irritation

Ambient