Please login to your online account to display your discounted pricing

Lead, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

For atomic absorption. (1mL = 1mg Pb) Solute: Lead Metal Solvent: Dilute Nitric Acid

$81.76 - $114.00

Chemical Identifiers

Chemical Name or Material Lead, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 10099-74-8
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134050
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SL21-100

 
fisher scientific company
SL21100
Poly Bottle 100mL Each for $81.76
Case of 6 Each for $490.56

SL21-500

 
fisher scientific company
SL21500
Poly Bottle 500mL Each for $114.00
Case of 6 Each for $684.00
Description & Specifications

Specifications

Purity Grade Certified Grade
Quantity 100mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Clear
Viscosity [NULL]
Chemical Name or Material Lead, Reference Standard Solution
Name Note 1000ppm ±1%
MDL Number MFCD00134050
ChemAlert Storage Symbol White
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
CAS 7697-37-2
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED Lead ref. Standard
Concentration 990 to 1010ppm
CAS 10099-74-8
CAS 7732-18-5
Safety and Handling