missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead Powder, -100 mesh, 99.95% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4560230

 View more versions of this product

Catalog No. AA4560230



Specifications

Specifications

Lead
7439-92-1
11.34g/cm3 at 20°C
Pb
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
Odorless
lead
5352425
Powder
250g
Gray to black atomized powder
99.95% (Metals basis)
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
[Pb]
207.2
CHEBI:27889
Black to Gray

Chemical Identifiers

7439-92-1
207.2
WABPQHHGFIMREM-UHFFFAOYSA-N
5352425
lead
Pb
MFCD00134050
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
CHEBI:27889
[Pb]
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 231-100-4

SDS Documents